Learn More
IL-17F Antibody (4H450) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marque:  Novus Biologicals NBP2-27319
Code nomenclature Nacres: NA.46
Code produit. 18717233
  • 585.00€ / 0.10mg
Description
IL-17F Monoclonal specifically detects IL-17F in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Monoclonal
Store at -20C. Avoid freeze-thaw cycles.
Purified
IgG1 κ