Learn More
IL-33 Rabbit anti-Mouse, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 28035-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The IL-33 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse IL-33. This antibody recognizes mouse, rat antigen. The IL-33 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
IL-33 | |
Polyclonal | |
Unconjugated | |
150 μL | |
77125 | |
IgG |
Western Blot, ELISA | |
200 μg/mL | |
Rabbit | |
Primary | |
Mouse, Rat |