Learn More
IL-33 Rabbit anti-Mouse, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 28035-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16835965
  • 325.38€ / 150µL
Expédition estimée: 28-09-2022
Description
The IL-33 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse IL-33. This antibody recognizes mouse, rat antigen. The IL-33 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Spécification
IL-33
Polyclonal
Unconjugated
150 μL
77125
IgG
ELISA, Western Blot
200 μg/mL
Rabbit
Primary
Rat, Mouse