Learn More
IL-6R Mouse anti-Human, Unconjugated, Clone: 2A5F4, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 66855-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16829115
  • 140.00€ / 20µL
Expédition estimée: 14-05-2024
Description
The IL-6R antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IL-6R. This antibody recognizes human antigen. The IL-6R antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
Spécification
IL-6R
Monoclonal
667 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
2A5F4
Unconjugated
20 μL
3570
IgG1