Learn More
IL-6R Mouse anti-Human, Unconjugated, Clone: 2A5F4, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66855-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The IL-6R antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IL-6R. This antibody recognizes human antigen. The IL-6R antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
IL-6R | |
Monoclonal | |
667 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
2A5F4 | |
Unconjugated | |
20 μL | |
3570 | |
IgG1 |