missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1R2 Mouse anti-Human, Mouse, Clone: 3H4H4, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 60262-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein binds interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA), and acts as a decoy receptor that inhibits the activity of its ligands. Interleukin 4 (IL4) is reported to antagonize the activity of interleukin 1 by inducing the expression and release of this cytokine. This gene and three other genes form a cytokine receptor gene cluster on chromosome 2q12. Two alternatively spliced transcript variants encoding the same protein have been reported.Spécification
IL1R2 | |
Monoclonal | |
2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P27930, P27931 | |
Il1r2 | |
IL1R2 Fusion Protein Ag4495 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG2a |
Immunofluorescence, Immunocytochemistry, Western Blot | |
3H4H4 | |
Unconjugated | |
IL1R2 | |
CD121b, CDw121b, IL 1R 2, IL 1R beta, IL 1RT 2, IL 1RT2, IL1 Receptor 2, IL-1 Receptor 2, IL1 Receptor II, IL-1 Receptor II, IL1R2, IL1RB, IL-1RII, Interleukin 1 receptor beta, Interleukin 1 receptor type 2, Interleukin 1 receptor type II | |
Mouse | |
Protein A | |
RUO | |
16178, 7850 | |
-20°C | |
Liquid |