missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL31 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22859-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Interleukin-31 (IL-31) is a recently discovered T-cell cytokine closely related to IL-6 type cytokines and is preferentially produced by T helper type 2 cells. IL-31 activity is mediated through the ligand-induced oligomerization of a dimeric receptor complex containing IL-31 receptor A and oncostatin M receptor. In response to IL-31 binding, these proteins activate the JAK/STAT and the AKT signaling pathways. RNA levels of IL-31 receptor A and oncostatin M receptor are induced in activated monocytes but are expressed constitutively in epithelial cells. IL-31, when overexpressed in transgenic mice, results in the development of pruritis, alopecia, and skin lesions and in humans may result in atopic dermatitis, suggesting that IL-31 may represent a novel target for antipruritic drug development.Spécification
IL31 | |
Polyclonal | |
Unconjugated | |
IL31 | |
IL 31, IL31, interleukin 31 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
386653, 76399 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6EAL8, Q6EBC2 | |
IL31 | |
IL31 Fusion Protein Ag18848 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |