missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EnquireBio IL3RA / CD123 (Acute Myeloid Leukemia Marker) Mouse, Clone: IL3RA/2065, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 3563-MSM3-P1
Informations supplémentaires : Poids : 0.01000kg
Spécification
IL3RA / CD123 (Acute Myeloid Leukemia Marker) | |
Cell Surface and Cytoplasmic | |
IL3RA/2065 | |
Unconjugated | |
Mouse | |
∽70kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
3563 | |
IgG2a κ |
ELISA | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human IL3RA protein fragment (apprx. aa 26-171) | |
THP-1, RPMI-8226 or HDLM-2 cells. Tonsil, Lymph Node or Stomach | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |