missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL3RA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13655-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
IL-3Ra (Interleukin-3 receptor subunit alpha) belongs to the type I cytokine receptor family and is a heterodimer with a unique alpha chain paired with the common beta (beta c or CDw131) subunit. IL-3Ra is responsible for the transmission of signal of Interleukin-3. IL-3Ra and IL-3Rb chain heterodimerization is induced by Interleukin-3, which is required for receptor activation but not high affinity binding. IL-3Ra is also reported that IL-3Ra subunits are overexpressed on acute myeloid leukemia (AML) blasts compared with normal hematopoietic cells and are thus potential targets for novel therapeutic agents.Spécification
IL3RA | |
Polyclonal | |
Unconjugated | |
IL3RA | |
CD123, hIL 3Ra, IL 3 receptor subunit alpha, IL 3R alpha, IL 3R subunit alpha, IL 3RA, IL3R, IL3RA, IL3RAY, IL3RX, IL3RY | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3563 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P26951 | |
IL3RA | |
IL3RA Fusion Protein Ag4452 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |