missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26677-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.Spécification
IL5 | |
Polyclonal | |
Unconjugated | |
IL5 | |
EDF, IL 5, IL5, Interleukin 5, T cell replacing factor, TRF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3567 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P05113 | |
IL5 | |
IL5 Fusion Protein Ag24583 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |