missing translation for 'onlineSavingsMsg'
Learn More

ILF3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 19887-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16829354

  • 325.38€ / 150µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

UBA6 (UBE1L2, MOP-4)is a novel E1 enzyme specific for ubiquitin and belongs to the ubiquitin-activating E1 family. It is a part of the cascade to attach ubiquitin (Ub) covalently to substrate proteins.UBA6 activates ubiquitin by first adenylating its carboxy-terminal glycine residue with ATP and then linking this residue to the side chain of a cysteine residue in E1, yielding an ubiquitin-E1 thioester and free AMP. Its isoforms influence spermatogenesis and male fertility.UBA6 is highly expressed in human monocytes.
Spécification

Spécification

ILF3
Polyclonal
Unconjugated
ILF3
CBTF, DRBF, DRBP76, ILF3, M phase phosphoprotein 4, MMP4, MPHOSPH4, MPP4, NF AT 90, NF110, NF90, NFAR, NFAR 1, NFAR2, TCP110, TCP80
Rabbit
Antigen Affinity Chromatography
RUO
16201, 3609, 84472
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q12906, Q9JIL3, Q9Z1X4
ILF3
ILF3 Fusion Protein Ag13707
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions