Learn More
IMP5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12664-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Intramembrane-cleaving aspartic protease (I-CLiP) that may be able to cleave type II membrane signal peptides in the hydrophobic plane of the membrane.Spécification
| IMP5 | |
| Polyclonal | |
| Unconjugated | |
| SPPL2C | |
| IMP 5, IMP5, intramembrane protease 5, SPP like 2C, SPPL2c | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 162540 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IUH8 | |
| SPPL2C | |
| IMP5 Fusion Protein Ag3345 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |