Learn More
INADL Mouse, anti-Human, Unconjugated, Clone: 3C6A4, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67833-1-IG-150UL
Informations supplémentaires : Poids : 0.00020kg
 Afficher plus de versions de ce produit
Code produit. 17281486
  • 349.00€ / 150µL
Expédition estimée: 18-10-2023
pour voir le stock disponible 
Description
Description
The INADL antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human INADL. This antibody recognizes human antigen. The INADL antibody has been validated for the following applications: WB,ELISA analysis.
Spécification
Spécification
INADL
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
3C6A4
Unconjugated
150 μL
10207