missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H3N2 Hemagglutinin Antibody - (A/Texas/1/77) - BSA Free, Novus Biologicals™
Goat Polyclonal Antibody
Marque: Novus Biologicals NB100-64556
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Influenza A H3N2 Hemagglutinin Polyclonal antibody specifically detects Influenza A H3N2 Hemagglutinin in Influenza A Virus H3N2 samples. It is validated for Immunocytochemistry, Immunofluorescence, Hemagglutination Assay.
Spécification
| Influenza A H3N2 Hemagglutinin | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.09% Sodium Azide | |
| Influenza A, Texas 1/77 strain (H3N2). | |
| 0.5 mL | |
| Virology Bacteria and Parasites | |
| Specific for Influenza A H3N2 by Haemagglutination inhibition. Does not react with influenza B, RSV, Para 1-3 or Adenovirus, but may react with proteins present on uninfected chicken cells. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Hemagglutination Assay | |
| 1.0 mg/mL | |
| Immunocytochemistry/Immunofluorescence 1:10-1:2000, Haemagglutination | |
| Goat | |
| IgG purified | |
| RUO | |
| Primary | |
| Influenza virus | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu