missing translation for 'onlineSavingsMsg'
Learn More

INPP5B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 15141-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16834194

  • 121.38€ / 20µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Cellular calcium signaling is controlled by the production of inositol phosphates (IPs) by phospholipase C in response to extracellular signals. The IP signaling molecules are inactivated by a family of inositol polyphosphate-5-phosphatases (5-phosphatases). INPP5B encodes the type II 5-phosphatase. The protein is localized to the cytosol and mitochondria, and associates with membranes through an isoprenyl modification near the C-terminus. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Spécification

Spécification

INPP5B
Polyclonal
Unconjugated
INPP5B
5PTase, INPP5B, Phosphoinositide 5 phosphatase
Rabbit
Antigen Affinity Chromatography
RUO
16330, 362590, 3633
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P32019, Q8K337
Inpp5b
INPP5B Fusion Protein Ag7315
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions