Learn More
INSL3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17997-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Seems to play a role in testicular function. May be a trophic hormone with a role in testicular descent in fetal life. Is a ligand for LGR8 receptor.Spécification
INSL3 | |
Polyclonal | |
Unconjugated | |
INSL3 | |
INSL3, Insulin like 3, insulin like 3 (Leydig cell), Ley I L, Leydig insulin like peptide, Relaxin like factor, RLF, RLNL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3640 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P51460 | |
INSL3 | |
INSL3 Fusion Protein Ag12455 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |