missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15848-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Insulin is a peptide hormone secreted by beta cells of the pancreatic islets. It regulates carbohydrate, protein and lipid metabolism by enhancing membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides and synthesis of proteins and nucleic acids. Insulin is 51-amino acid polypeptide product produced from a precursor peptide, proinsulin. Proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds and one molecule of C-peptide. Insulin deficiency results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of b-cell origin such as insulinoma.Spécification
Insulin | |
Polyclonal | |
Unconjugated | |
INS | |
ILPR, INS, insulin, IRDN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3630 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P01308 | |
INS | |
Insulin Fusion Protein Ag8630 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |