missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin beta-6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21703-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ITGB6 (Integrin alpha-V/beta-6) is a receptor for fibronectin and cytotactin. It recognizes the sequence R-G-D in its ligands. Internalization of integrin alpha-V/beta-6 via clathrin-mediated endocytosis promotes carcinoma cell invasion.Spécification
Integrin beta-6 | |
Polyclonal | |
Unconjugated | |
ITGB6 | |
Integrin beta 6, integrin, beta 6, ITGB6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16420, 311061, 3694 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P18564, Q6AYF4, Q9Z0T9 | |
Itgb6 | |
Integrin beta-6 Fusion Protein Ag16319 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |