Learn More
Thermo Scientific™ IOPQ Qualification Service, Documentation and 1 Temp Mapping for IntelliStack Incubator Shaker
IOPQ Qualification Service, Documentation and 1 Temp Mapping for IntelliStack Incubator Shaker
Marque: Thermo Scientific™ IOQPT3510
Description
- IQ: Verifies that the installation site and equipment environment meet manufacturer-specified environmental requirements. Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment and any other accessories are assembled and installed. Configuration information for each system component gets recorded.
- Installation of equipment is NOT included
- OQ: Verifies important equipment functions and ensures that the equipment operates as expected by the manufacturer and in conformance to standards and requirements.
- Key tests: Door Ajar, Power Failure, Temperature Alerts
Temp Mapping service will include:
- 1 open door temperature recovery test
- 1 power failure test
- Min and max values along with averages
- Temperature graph over a time period
- Detailed reports will include raw probe data
All of this will be performed and delivered by a Thermo Fisher Qualified Engineer equipped with training certificates, temperature mapping data logger calibrated annually with metrology certificates.
Note: Study requirements greater than 24 hours will be reviewed and quoted
Spécification
IQ/OQ/PQ Service |
Suggestions de produits
Customers who viewed this item also viewed.