Learn More
IPO4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 11679-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16828923
  • 399.00€ / 150µL
Expédition estimée: 15-04-2024
Description
IPO4 gene ontology annotations related to this gene include protein import into nucleus.
Spécification
Spécification
IPO4
Polyclonal
Unconjugated
IPO4
Imp4, Imp4b, importin 4, Importin 4b, IPO4, Ran binding protein 4, RanBP4
Rabbit
Antigen Affinity Chromatography
RUO
290228, 75751, 79711
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8TEX9, Q8VI75
Ipo4
IPO4 Fusion Protein Ag2281
150 μL
Primary
Human, Mouse, Rat