missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IQGAP3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25930-1-AP-150UL
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
IQGAP3 (IQ motif containing GTPase activating protein 3) is a 1, 631 amino acid protein that acts as an effector of Cdc42 and Rac 1, linking their activation to the cytoskeleton during neuronal morphogenesis. A novel member of the IQGAP family, IQGAP3 is highly expressed in brain where it localizes to axons of hippocampal neurons. IQGAP3 contains one Ras-GAP domain, a CH (calponin-homology) domain, four IQ domains and is encoded by a gene located on human chromosome 1, which spans 260 million base pairs, contains over 3, 000 genes and comprises nearly 8% of the human genome. Chromosome 1 houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson′s disease, Gaucher disease, schizophrenia and Usher syndrome. Aberrations in chromosome 1 are found in a variety of cancers, including head and neck cancer, maligt melanoma and multiple myeloma.Spécification
IQGAP3 | |
Polyclonal | |
Unconjugated | |
IQGAP3 | |
IQGAP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
128239 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86VI3 | |
IQGAP3 | |
IQGAP3 Fusion Protein Ag23109 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |