missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRAK1 Mouse anti-Human, Clone: 1H10A7, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66653-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Interleukin-1 receptor-associated kinase 1 (IRAK1) is a serine/threonine-protein kinase that plays a critical role in initiating innate immune response against foreign pathogens. IRAK is associated with Toll-like receptor (TLR) and IL-1R signaling pathways. It is rapidly recruited by MYD88 to the receptor-signaling complex upon TLR activation. IRAK′s association with MYD88 leads to IRAK1 phosphorylation by IRAK4 and subsequent autophosphorylation and kinase activation. Alternatively, IRAK1 can phosphorylate TIRAP to promote its ubiquitination and subsequent degradation. IRAK1 also phosphorylates the interferon regulatory factor 7 (IRF7) to induce its activation and translocation to the nucleus. This results in the transcriptional activiation of type I IFN genes, which drive the cell in an antiviral state.Spécification
IRAK1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P51617 | |
IRAK1 | |
IRAK1 Fusion Protein Ag0728 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
1H10A7 | |
Unconjugated | |
IRAK1 | |
IRAK, IRAK 1, IRAK1, pelle | |
Mouse | |
Protein A | |
RUO | |
3654 | |
-20°C | |
Liquid |