missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JNK1 Antibody (S05-7A3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19929-50ul
This item is not returnable.
View return policy
Description
JNK1 Monoclonal antibody specifically detects JNK1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| JNK1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S05-7A3 | |
| Unconjugated | |
| c-Jun N-terminal kinase 1, EC 2.7.11, EC 2.7.11.24, JNK1 alpha protein kinase, JNK1 beta protein kinase, JNK1JNK1A2, JNK-46, JUN N-terminal kinase, MAP kinase 8, MAPK 8, mitogen-activated protein kinase 8, mitogen-activated protein kinase 8 isoform JNK1 alpha1, mitogen-activated protein kinase 8 isoform JNK1 beta2, PRKM8JNK, protein kinase JNK1, SAPK1JNK21B1/2, Stress-activated protein kinase 1, Stress-activated protein kinase JNK1 | |
| Recombinant protein of human JNK1 (Uniprot # Q91Y86) | |
| 50 μg | |
| Cancer, Hypoxia, Immunology, Innate Immunity, MAP Kinase Signaling, p53 Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| 5599 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction