missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL15 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25066-1-AP-150UL
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
KLHL15 is a member of a growing superfamily of kelch repeat-containing proteins. The kelch motif was discovered in the sequence of the Drosophila kelch ORF1 protein and forms a conserved beta-propeller tertiary structure. These kelch-like proteins have been implicated in embryogenesis and carcinogenesis through cytoskeleton organization. KLHL15 mRNA is widely expressed in many tissues and is thought to be involved in protein ubiquitination and cytoskeleton organization.Spécification
KLHL15 | |
Polyclonal | |
Unconjugated | |
KLHL15 | |
kelch like 15 (Drosophila), Kelch like protein 15, KIAA1677, KLHL15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80311 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96M94 | |
KLHL15 | |
KLHL15 Fusion Protein Ag16878 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |