Learn More
KLRG1 Monoclonal Antibody (2F1), Alexa Fluor™ 647, eBioscience™, Invitrogen™
Syrian Hamster Monoclonal Antibody
Marque: Affymetrix eBioscience 51-5893-80
Informations supplémentaires : Poids : 0.01000kg
Description
Description: This 2F1 monoclonal antibody reacts with the mouse Killer cell Lectin-like Receptor G1 (KLRG1), also known as Mast cell Function-associated Antigen (MAFA). KLRG1 is a homodimer of glycosylated 30-38 kDa subunits and contains a cytoplasmic motif similar to the immunoreceptor tyrosine-based inhibitory motif (ITIM). Rat MAFA was identified as an antigen specific to rat mast cells; however, the expression of mouse KLRG1/MAFA using 2F1 has not been detected on the surface of mouse mast cell lines, bone marrow-derived mast cells, or peritoneal mast cells. This antigen is expressed on approximately one-third of mouse NK cells and a subset of T cells. MHC class I molecules regulate KLRG1 via interactions with class I-specific inhibitory Ly49 molecules and SHP-1 signaling. Although KLRG1 and Ly49 are both lectin-like inhibitory receptors that are regulated by class I MHC expression, the effects of this on cell surface expression of these molecules are opposing, and the underlying regulatory mechanisms distinct. Applications Reported: This 2F1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2F1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.
Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. KLRG1 (Killer cell lectin-like receptor G1, MAFA, 2F1-Ag) is a homodimeric member of the lectin-like type 2 transmembrane receptor family whose members contain characteristic immunoreceptor tyrosine-based inhibitory motifs (ITIMs) in their cytoplasmic domains. These ITIMs interact with the SH2 domains of protein phosphatases such as SHP-1. KLRG1 is an inhibitory receptor that is expressed on natural killer (NK) cells and certain T cells. NK cells are involved in the lysis of tumor cells and virus-infected cells, and mediate humoral and cell-mediated immune responses.Spécification
KLRG1 | |
Monoclonal | |
0.2 mg/ml | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
O88713 | |
Klrg1 | |
Affinity chromatography | |
RUO | |
50928 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
2F1 | |
Alexa Fluor 647 | |
Klrg1 | |
Klrg1 | |
Syrian Hamster | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |