missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRG1 Mouse anti-Human, RY586, Clone: Z7-205.rMAb, BD Horizon™
Mouse Monoclonal Antibody
Marque: BD Biosciences 568493
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
KLRG1 Monoclonal antibody specifically detects KLRG1 in Human samples. It is validated for Flow cytometry (Routinely Tested)
Spécification
| KLRG1 | |
| Monoclonal | |
| RY586 | |
| Killer cell lectin-like receptor subfamily G member 1; MAFA | |
| Hepa 1-6 cells transfected with hKLRG1 DNA | |
| 100 Tests | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| Z7-205.rMAb | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10219 | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
| IgG1 κ |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu