missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRG1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10974-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Killer cell lectin-like receptor G1 (KLRG1; formerly known as mouse MAFA or 2F1-Ag), is a homodimeric member of the lectin-like type 2 transmembrane receptor family whose members contain characteristic immunoreceptor tyrosine-based inhibitory motifs (ITIMs) in their cytoplasmic domains. These ITIMs interact with the SH2 domains of protein phosphatases such as SHP-1.Spécification
| KLRG1 | |
| Polyclonal | |
| Unconjugated | |
| KLRG1 | |
| 2F1, CLEC15A, KLRG1, MAFA, MAFA 2F1, MAFA L, MAFA LIKE, MAFA like receptor, MAFAL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10219 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96E93 | |
| KLRG1 | |
| KLRG1 Fusion Protein Ag1407 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu