missing translation for 'onlineSavingsMsg'
Learn More

KRT40 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 25398-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16833975

  • 325.38€ / 150µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Cytokeratins, a group comprising at least 29 different proteins, are characteristic of epithelial and trichocytic cells. Cytokeratin 5, 6 and 8 are members of the type II neutral-to-basic subfamily. Monoclonal antibodies to cytokeratins are specific markers of epithelial cell differentiation and have been widely used as tools in tumor identification and classification. PCK-26 facilitates the typing of normal, metaplastic and neoplastic cells and it may aid in the discrimination of carcinomas and non-epithelial tumors such as sarcomas, lymphomas and neural tumors. It is also useful in detecting micro-metastases in lymph nodes and other tissues and for determining the origin of poorly differentiated tumors.
Spécification

Spécification

KRT40
Polyclonal
Unconjugated
KRT40
CK 40, Cytokeratin 40, K40, KA36, keratin 40, KRT40, Type I hair keratin Ka36
Rabbit
Antigen Affinity Chromatography
RUO
125115
-20°C
Liquid
Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6A162
KRT40
KRT40 Fusion Protein Ag21889
150 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions