missing translation for 'onlineSavingsMsg'
Learn More

L-Plastin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 13025-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16831034

  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

L-plastin is a leukocyte specific actin bundling protein from fimbrin family of proteins. It has two actin-binding domains and two EF hand type calcium-binding domains. It gets phosphorylated on the serine residue in its headpiece region in response to a variety of inflammatory stimuli. It is hypothesized that L-plastin may play a role in regulating integrin mediated adhesion in leukocytes. L-plastin is expressed in majority of human cancer cell lines derived from solid tumors. Its expression is positively upregulated by testosteronse in andrgen receptor positive prostrate and breast cancer cells through three androgen receptor binding elements located in the upstream region of the gene.
Spécification

Spécification

L-Plastin
Polyclonal
Unconjugated
LCP1
L PLASTIN, LCP 1, LCP1, Lymphocyte cytosolic protein 1, Plastin 2, PLS2
Rabbit
Antigen Affinity Chromatography
RUO
18826, 306071, 3936
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Flow Cytometry
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P13796, Q61233
Lcp1
L-Plastin Fusion Protein Ag3571
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions