missing translation for 'onlineSavingsMsg'
Learn More

CD15 Ab-3 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry and flow cytometry procedures with Epredia™ CD15 Ab-3, Mouse Monoclonal Antibody.

Marque:  Epredia MS-1259-P0

Code nomenclature Nacres: NA.41

Informations supplémentaires : Poids : 0.00170kg

 Afficher plus de versions de ce produit

Code produit. 12689396

  • 129.00€ / 100µL
Expédition estimée: 06-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

3-fucosyl-N-acetyllactosamine (3-FAL) or CD15 or X-hapten plays a role in mediating phagocytosis, bactericidal activity, and chemotaxis. It is present on >95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. CD15 is also expressed in Reed-Sternberg cells and some epithelial cells. CD15 antibody is very useful in the identification of Hodgkin's disease. CD15 is occasionally expressed in large cell lymphomas of both B and T phenotypes which otherwise have a quite distinct histological appearance.

Host Species: Mouse

Clone: MMA; same as LeuM1

Isotype: IgM/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: U937 histiocytic cell line

Molecular Weight: 220kDa

Positive Control: Reed-Sternberg's cells in Hodgkin's lymphoma

Cellular Localization: Membrane and granular paranuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry
Spécification

Spécification

CD15 Ab-3
Monoclonal
200μg/mL
Mouse
100 μL
Oncogenesis
Human
IgM κ
Flow Cytometry, Immunohistochemistry (Paraffin)
MMA; same as LeuM1
Unconjugated
U937 histiocytic cell line
IVD
Primary
Concentrated
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions