Learn More
CD99/MIC2 (Ewing’s Sarcoma Marker) Ab-2 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry applications with Epredia™ CD99/MIC2 (Ewing's Sarcoma Marker) Ab-2, Mouse Monoclonal Antibody.
Marque: Epredia MS-1633-P1ABX
Description
CD99, or MIC2 gene product, or E2 antigen is expressed on the cell membrane of some lymphocytes, cortical thymocytes, and granulosa cells of the ovary. The antigen is also expressed by most pancreatic islet cells, Sertoli cells of the testis, and some endothelial cells. Mature granulocytes express very little or no CD99. MIC2 is strongly expressed on Ewing's sarcoma cells and primitive peripheral neuroectodermal tumors.Host Species: Mouse
Clone: O13
Isotype: IgG1
Species Reactivity: Human. Does not react with mouse and rat. Others not known.
Epitope: Not determined
Immunogen: MeWo human melanoma cells
Molecular Weight: 27-32 kDa
Positive Control: Pancreas or Ewing's sarcoma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Spécification
| CD99/MIC2 (Ewings Sarcoma Marker) Ab-2 | |
| Monoclonal | |
| 1mg/mL | |
| Mouse | |
| 100 μL | |
| Oncogenesis | |
| Human | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| O13 | |
| Unconjugated | |
| MeWo human melanoma cells | |
| RUO | |
| Primary | |
| Concentrated |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.