Learn More
Kappa Light Chain Ab-1 Mouse Monoclonal Antibody, Epredia™
Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Kappa Light Chain Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
Marque:  Epredia MS-143-P0
Code nomenclature Nacres: NA.46
Code produit. 12619806
Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
Voir les produits de remplacement »
Description
Description
Antibody to the κ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
Host Species: Mouse
Clone: L1C1
Isotype: IgG1/k
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: Not determined
Immunogen: B-lymphoma cells
Molecular Weight: 25kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Kappa Light Chain Ab-1
Monoclonal
200μg/mL
Mouse
100 μL
Hematopoietic
Human
IgG2a κ
Immunohistochemistry (Paraffin)
L1C1
Unconjugated
B lymphoma cells
IVD
Primary
Concentrated