missing translation for 'onlineSavingsMsg'
Learn More

LACTB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 18195-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16828254

  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody is specific to LACTB.

This gene encodes a protein from the large 39S subunit of the mitochondrial ribosome (mitoribosome). The encoded protein has some sequence similarity to prokaryotic beta-lactamases but most of the residues that are responsible for the beta-lactamase activity are not conserved between the two proteins.
Spécification

Spécification

LACTB
Polyclonal
Unconjugated
LACTB
G24, lactamase, beta, LACTB, MRPL56
Rabbit
Antigen Affinity Chromatography
RUO
114294, 300803, 80907
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P83111, Q9EP89
Lactb
LACTB Fusion Protein Ag12233
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions