missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LENG9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16295-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
LENG9 | |
Polyclonal | |
Unconjugated | |
LENG9 | |
LENG9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
94059 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96B70 | |
LENG9 | |
LENG9 Fusion Protein Ag9481 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |