Learn More
LIG1 Mouse, anti-Human, Unconjugated, Clone: 3D9G8, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67840-1-IG-150UL
Informations supplémentaires : Poids : 0.00020kg
Code produit. 17281496
  • 349.00€ / 150µL
Expédition estimée: 09-10-2023
Description
LIG1, also named as LRIG1, acts as a feedback negative regulator of signaling by receptor tyrosine kinases, through a mechanism that involves enhancement of receptor ubiquitination and accelerated intracellular degradation.
Spécification
Spécification
LIG1
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rat
Western Blot, ELISA
3D9G8
Unconjugated
150 μL
3978