missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMOD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15117-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The leiomodin 1 protein (LMOD1) has a putative membrane-spanning region and 2 types of tandemly repeated blocks. The transcript is expressed in all tissues tested, with the highest levels in thyroid, eye muscle, skeletal muscle, and ovary. Increased expression of leiomodin 1 may be linked to Graves′ disease and thyroid-associated ophthalmopathy. The leiomodin 1 protein has a putative membrane-spanning region and 2 types of tandemly repeated blocks. The transcript is expressed in all tissues tested, with the highest levels in thyroid, eye muscle, skeletal muscle, and ovary. Increased expression of leiomodin 1 may be linked to Graves′ disease and thyroid-associated ophthalmopathy.Spécification
LMOD1 | |
Polyclonal | |
Unconjugated | |
LMOD1 | |
1D, 64 kDa autoantigen 1D, 64 kDa autoantigen 1D3, 64 kDa autoantigen D1, 64kD, D1, Leiomodin 1, leiomodin 1 (smooth muscle), Leiomodin, muscle form, LMOD1, SM LMOD, Smooth muscle leiomodin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25802, 304816, 93689 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A0A0G2K0D3, P29536, Q8BVA4 | |
Lmod1 | |
LMOD1 Fusion Protein Ag7176 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |