missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC38 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 55479-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Auxiliary protein of the large-conductance, voltage and calcium-activated potassium channel (BK alpha). Modulates gating properties by producing a marked shift in the BK channel′s voltage dependence of activation in the hyperpolarizing direction, and in the absence of calcium.Spécification
LRRC38 | |
Polyclonal | |
Unconjugated | |
LRRC38 | |
LRRC38, RP4 597A16.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
126755, 242735 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2VDH3, Q5VT99 | |
Lrrc38 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |