Learn More
Ly-6G Rat, FITC, Clone: RB6-8C5, enQuireBio™
Rat Monoclonal Antibody
Marque:  EnquireBio QAB77-F-500ug
Code nomenclature Nacres: NA.46
Code produit. 15982458
  • 327.60€ / 500µg
Spécification
Ly-6G
Monoclonal
0.2mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Ly6c1
Protein A/G Purified
RUO
17067
IgG2b κ
Flow Cytometry
RB6-8C5
FITC
NP_001238984
Rat
500 μg
Primary
Store at 2-4°C.