missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYRM7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23833-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
LYRM7 | |
Polyclonal | |
Unconjugated | |
LYRM7 | |
C5orf31, LYR motif containing protein 7, LYRM7, Lyrm7 homolog (mouse) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
90624 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5U5X0 | |
LYRM7 | |
LYRM7 Fusion Protein Ag20696 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |