missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mammaglobin (SCGB2A2) (Breast Cancer Marker) Mouse, Clone: MGB1/2000, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 4250-MSM1-P1
Informations supplémentaires : Poids : 0.01000kg
Spécification
Mammaglobin (SCGB2A2) (Breast Cancer Marker) | |
Cytoplasmic, Cell Surface and Nuclear | |
MGB1/2000 | |
Unconjugated | |
Mouse | |
18kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
4250 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human Mammaglobin (SCGB2A2) protein | |
SK-BR-3 cells or Breast Carcinoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |