Learn More
MCL1 Mouse anti-Human, Clone: 1E3C2, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66026-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
YY1 is a ubiquitously distributed transcription factor belonging to the GLI-Kruppel class of zinc finger proteins. The protein is involved in repressing and activating a diverse number of promoters. YY1 may direct histone deacetylases and histone acetyltransferases to a promoter in order to activate or repress the promoter, thus implicating histone modification in the function of YY1.Spécification
MCL1 | |
Monoclonal | |
1.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q07820 | |
MCL1 | |
MCL1 Fusion Protein Ag10609 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
1E3C2 | |
Unconjugated | |
MCL1 | |
EAT, Mcl 1, Mcl 1l, Mcl 1s Delta TM, MCL1, mcl1/EAT, MCL1L, MCL1S, TM | |
Mouse | |
Protein A | |
RUO | |
4170 | |
-20°C | |
Liquid |