missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mcp1 Mouse anti-Mouse, Clone: 1B9F7, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66272-1-IG-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This chemotactic factor attracts monocytes and basophils but not neutrophils or eosinophils and augments monocyte anti-tumor activity. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis or atherosclerosis. It may be involved in the recruitment of monocytes into the arterial wall during the disease process of atherosclerosis.Spécification
| Mcp1 | |
| Monoclonal | |
| 1.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P10148 | |
| Ccl2 | |
| Mcp1 Fusion Protein Ag24085 | |
| 150 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1B9F7 | |
| Unconjugated | |
| Ccl2 | |
| AI323594, Ccl2, chemokine (C C motif) ligand 2, HC11, JE, MCAF, MCP 1, Mcp1, Scya2, Sigje, SMC CF | |
| Mouse | |
| Protein G | |
| RUO | |
| 20296 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mcp1 Mouse anti-Mouse, Clone: 1B9F7, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction


