Learn More
MCT2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 20355-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16829884
  • 325.38€ / 150µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 
Description
Description
The MCT2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human MCT2. This antibody recognizes human, mouse antigen. The MCT2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
Spécification
MCT2
Polyclonal
Rabbit
Primary
Mouse, Human
Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
150 μL
9194