missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGAT4A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25109-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a key glycosyltransferase that regulates the formation of tri- and multiantennary branching structures in the Golgi apparatus. The encoded protein, in addition to the related isoenzyme B, catalyzes the transfer of N-acetylglucosamine (GlcNAc) from UDP-GlcNAc in a beta-1,4 linkage to the Man-alpha-1,3-Man-beta-1,4-GlcNAc arm of R-Man-alpha-1,6(GlcNAc-beta-1,2-Man-alpha-1,3)Man-beta-1,4-GlcNAc-beta-1,4-GlcNAc-beta-1-Asn. The encoded protein may play a role in regulating the availability of serum glycoproteins, oncogenesis, and differentiation.Spécification
MGAT4A | |
Polyclonal | |
Unconjugated | |
MGAT4A | |
GlcNAc T IVa, GNT IV, GNT IVA, MGAT4A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11320 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UM21 | |
MGAT4A | |
MGAT4A Fusion Protein Ag18956 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |