Learn More
MHC Class II (I-A/I-E) Monoclonal Antibody (M5/114.15.2), Super Bright™ 600, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
178.00€ - 362.00€
Spécification
Antigène | MHC Class II (I-A/I-E) |
---|---|
Clone | M5/114.15.2 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15983544
|
Affymetrix eBioscience
63-5321-80 |
25 μg |
178.00€
25µg |
En stock Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15993544
|
Affymetrix eBioscience
63-5321-82 |
100 μg |
362.00€
100µg |
Expédition estimée: 15-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Description: The M5/114.15.2 monoclonal antibody reacts with the mouse major histocompatibility complex class II, both I-A and I-E subregion-encoded glycoproteins (I-A b, I-A d, I-A q, I-E d, I-E k, not I-A f, I-A k, or I-A s). It detects a polymorphic determit present on B cells, monocytes, macrophages, dendritic cells, and activated T lymphocytes from mice carrying the H-2 b, H-2 d, H-2 q, H-2 p, H-2 r and H-2 u but not from mice carrying the H-2 s or H-2 f haplotypes. The M5/114 mAb is reported to inhibit I-A-restricted T cell responses of the H-2 b, H-2 d, H-2 q, H-2 u but not H-2 f, H-2 k, or H-2 s haplotypes. Applications Reported: This M5/114.15.2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This M5/114.15.2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 600 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 600 nm. We recommend using a 610/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome.
MHC Class II (I-A/I-E) Monoclonal antibody specifically detects MHC Class II (I-A/I-E) in Mouse samples. It is validated for Flow Cytometry.Spécification
MHC Class II (I-A/I-E) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Abeta; AI323765; AI845868; cell surface glycoprotein; E-alpha-f; H-2 class II histocompatibility antigen, A beta chain; H-2 class II histocompatibility antigen, E-D alpha chain; H-2 class II histocompatibility antigen, E-K alpha chain; H-2 class II histocompatibility antigen, E-U alpha chain; H-2 class II histocompatibility antigen, I-E beta chain; H-2Ab; H2-Ab; H2-Ab1; H-2Ea; H2-Ea; H2-Ea-ps; H2Eb; H-2Eb; H2-Eb1; H2-iabeta; H2-IE-alpha; histocompatibility 2, class II antigen A, beta 1; histocompatibility 2, class II antigen E alpha; histocompatibility 2, class II antigen E alpha, pseudogene; histocompatibility 2, class II antigen E beta; Ia2; Ia-2; Ia3; Ia-3; Ia4; Ia-4; IAb; I-Ab; I-Abeta; I-Ad; I-Aq; I-E alpha MHC class II; I-E beta MHC class II; I-Ealpha; I-Ed; I-Ek; major histocompatibility complex class II beta chain; major histocompatibility complex H-2E beta chain; MHC class 2; MHC class II antigen A beta; MHC class II antigen E alpha; MHC class II H2-IA-beta-psi; MHC class II I | |
H2-Ab1, H2-Eb1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
H2-Ab1 |
M5/114.15.2 | |
Flow Cytometry | |
Super Bright 600 | |
Rat | |
Mouse | |
P01921, P04230, P06342, P06343, P06344, P06345, P06346, P14483 | |
14961, 14969 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |