Learn More
Mind Bomb 1/MIB1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Marque: Novus Biologicals NBP3-11304-0.025mg
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Mind Bomb 1/MIB1 Polyclonal antibody specifically detects Mind Bomb 1/MIB1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Spécification
| Mind Bomb 1/MIB1 | |
| Polyclonal | |
| Western Blot 0.5 - 1.0 mg/mL, ELISA 1:16000, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Q86YT6 | |
| Goat | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Tris buffered saline, 0.5% BSA | |
| DAPK-interacting protein 1, DIP1, DIP-1FLJ90676, DKFZp686I0769, DKFZp761M1710, E3 ubiquitin-protein ligase MIB1, EC 6.3.2, EC 6.3.2.-, KIAA1323, MGC129660, MIB, Mind bomb homolog 1, mindbomb homolog 1 (Drosophila), ubiquitin ligase mind bomb, ubiquitin ligase protein MIB1, Zinc finger ZZ type with ankyrin repeat domain protein 2, ZZANK2MGC129659, ZZZ6 | |
| Synthetic peptide sequence C-DKDNTNVNADVQKLQ from the internal region of MIB1 (NP_065825.1). | |
| 0.025 mg | |
| Lipid and Metabolism, Zinc Finger | |
| 57534 | |
| Store at 4°C. | |
| IgG |