missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLH3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25298-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene is a member of the MutL-homolog (MLH) family of DNA mismatch repair (MMR) genes. MLH genes are implicated in maintaining genomic integrity during DNA replication and after meiotic recombination. The protein encoded by this gene functions as a heterodimer with other family members. Somatic mutations in this gene frequently occur in tumors exhibiting microsatellite instability, and germline mutations have been linked to hereditary nonpolyposis colorectal cancer type 7 (HNPCC7). Several alternatively spliced transcript variants have been identified, but the full-length nature of only two transcript variants has been determined.Spécification
MLH3 | |
Polyclonal | |
Unconjugated | |
MLH3 | |
HNPCC7, MLH3, mutL homolog 3 (E. coli), MutL protein homolog 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27030 | |
-20°C | |
Liquid |
Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UHC1 | |
MLH3 | |
MLH3 Fusion Protein Ag18201 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |