missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMACHC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21462-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The exact function of the protein encoded by this gene is not known, however, its C-terminal region shows similarity to TonB, a bacterial protein involved in energy transduction for cobalamin (vitamin B12) uptake. Hence, it is postulated that this protein may have a role in the binding and intracellular trafficking of cobalamin. Mutations in this gene are associated with methylmalonic aciduria and homocystinuria type cblC.Spécification
MMACHC | |
Polyclonal | |
Unconjugated | |
MMACHC | |
cblC, MMACHC, RP11 291L19.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25974, 313520, 67096 | |
-20°C | |
Liquid |
Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CZD0, Q9Y4U1 | |
MMACHC | |
MMACHC Fusion Protein Ag15001 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |