missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MME/CD10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | MME/CD10 |
---|---|
Concentration | 0.45 mg/mL |
Applications | Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16802515
|
Proteintech
23898-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 27-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16892505
|
Proteintech
23898-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 27-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
CD10 (Common Acute Lymphocytic Leukemia Antigen,CALLA), is a cell surface enzyme with neutral metalloendopeptidase activity which inactivates a variety of biologically active peptides. CD10 is expressed on the cells of lymphoblastic, Burkitt′s, and follicular germinal center lymphomas, immature B cells with in adult bone marrow and on cells from patients with chronic myelocytic leukemia (CML). CD10 is also present on breast myoepithelial cells, bile canaliculi, fibroblasts, with especially high expression on the brush border of kidney and gut epithelial cells. CD10 is a neutral endopeptidase that cleaves peptides at the amino side of hydrophobic residues and inactivates several peptide hormones including glucagon, enkephalins, substance P, neurotensin, oxytocin, and bradykinin. Further, CD10 is a 100 kDa type II transmembrane glycoprotein that exists in a single copy of greater than 45 kb. The 5′ untranslated region of the CD10 gene is alternatively spliced, resulting in four separate mRNA transcripts and the coding region is not affected by alternative splicing. Diseases associated with CD10 dysfunction include spinocerebellar ataxia 43 and Charcot-Marie tooth Disease.Spécification
MME/CD10 | |
Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
P07861, P08473, Q61391 | |
17380, 24590, 4311 | |
MME,CD10 Fusion Protein Ag20963 | |
Primary | |
-20°C | |
MME |
0.45 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Atriopeptidase, CALLA, CD10, Enkephalinase, EPN, membrane metallo endopeptidase, MME, NEP, Neprilysin, Neutral endopeptidase, Neutral endopeptidase 24.11 | |
Mme | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |