Learn More
MMP10 Recombinant Rabbit Monoclonal Antibody (24 H24L21), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen 701288
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
This antibody is predicted to react with mouse, rat, non-human primate and rabbit based on sequence homology. Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
MMP10 Recombinant Monoclonal antibody specifically detects MMP10 in Human, Rat samples. It is validated for Western Blot.Spécification
MMP10 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
AV377895; matrix metallo protease; matrix metallopeptidase 10; matrix metallopeptidase 10 (stromelysin 2); matrix metalloprotease 10; matrix metalloproteinase 10; matrix metalloproteinase 10 (stromelysin 2); matrix metalloproteinase-10; MMP; Mmp10; MMP-10; MMPs; SL-2; STMY2; stromelysin 2; stromelysin-2; Transformation-associated protein 34A; transin 2; transin-2 | |
Recombinant protein corresponding to amino acids 282–476 of human MMP10 | |
100 μg | |
Primary | |
Human, Rat | |
Liquid |
Western Blot | |
24 H24L21 | |
Unconjugated | |
P07152, P09238 | |
Rabbit | |
Protein A | |
RUO | |
117061, 4319 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |