missing translation for 'onlineSavingsMsg'
Learn More

MMP13 Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 18165-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16828224

  • 325.38€ / 150µL
Expédition estimée: 29-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

MMP13 (collagense 3) plays a role in the degradation of extracellular matrix proteins including fibrillar collagen, fibronectin, TNC, and ACAN. MMP13 cleaves tripal helical collagens, including type I, II, and III collagen - but has the highest activity with soluble type II collagen. It can degrade collagen type IV, XIV, and X. MMP13 plays a role in wound healing, tissue remodeling, cartilage degradation, bone development, bone mineralization, and ossification. It is required for normal embryonic bone development and ossificaiton. Mutations affecting the gene can lead to spondyloepimetaphyseal dysplasia Missouri type, metaphyseal anadysplasia 1, and metaphyseal dysplasia Sphar type.
Spécification

Spécification

MMP13
Polyclonal
Unconjugated
MMP13
CLG3, Collagenase 3, Matrix metalloproteinase 13, MMP 13, MMP13
Rabbit
Antigen Affinity Chromatography
RUO
171052, 17386, 387293, 4322
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P23097, P33435, P45452
Mmp13, mmp13a
MMP13 Fusion Protein Ag12653
150 μL
Primary
Zebrafish, Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions